Potassium chromate 1/30 M K2CrO4 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35157-1L
BJ35157-1LEA 178.91 USD
BJ35157-1L
Potassium chromate 1/30 M K2CrO4 (0.1N) volumetric solution, Fluka™
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6

Order Now

Specification Test Results

Factor 0.999 - 1.001

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR