Potassium hydroxide 1 M KOH (1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35113-1L
BJ35113-1LEA 207 USD
BJ35113-1L
Potassium hydroxide 1 M KOH (1N) volumetric solution, Fluka™
Potassium hydroxide
Formula: KOH
Boiling Pt: 100 °C
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR