Ammonium thiocyanate 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

35036-1L
BJ35036-1LEA 46.63 USD
BJ35036-1L
Ammonium thiocyanate 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™
Ammonium thiocyanate
Formula: NH₄SCN
Boiling Pt: 100 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR