Hydrogen chloride 0.1 M HCl (0.1N) in 2-propanol volumetric solution, for total base number, according to DIN 51558-1, Fluka™

Supplier: Honeywell Research Chemicals
Danger

35009-1L
BJ35009-1LEA 70.31 USD
BJ35009-1L
Hydrogen chloride 0.1 M HCl (0.1N) in 2-propanol volumetric solution, for total base number, according to DIN 51558-1, Fluka™
Hydrogen chloride
Formula: HCl
Boiling Pt: 82 °C
Melting Pt: no data available
Density: 0.788 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 200-661-7
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 0.099 - 0.101 mol/l
Water (Karl Fischer) Max. 1 %
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR