Bromate-Bromide 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
35007-1L
BJ35007-1LEA
131.88
USD
BJ35007-1L
Bromate-Bromide 0.25 M Br2 (0.5 N) volumetric solution, Fluka™
Bromate-Bromide
Titrant for the Determination of the Bromine Number
Specification Test Results
Factor | 0.998 - 1.002 |