Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
35006-1L
BJ35006-1LEA
124.73
USD
BJ35006-1L
Bromate-Bromide 0.05 M Br2 (0.1 N) volumetric solution, Fluka™
Bromate-Bromide
Titrant for the Determination of the Bromine Number
Specification Test Results
Factor | 0.998 - 1.002 |