Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

34978-4X4L
BJ34978-4X4LCS 675 USD
BJ34978-4X4L
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Water with 0,1% (v/v) trifluoroacetic acid

RTECS Number: 231-791-2

Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9982 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 76-05-1
EINECS: 200-929-3

Order Now

Specification Test Results

Assay trifluoroacetic acid 0.093 - 0.107 %
pH (20 °C) 2.0 - 2.2
Calcium (Ca) max. 0.5 ppm
Potassium (K) max. 0.5 ppm
Magnesium (Mg) max. 0.5 ppm
Sodium (Na) max. 2 ppm
Transmittance at 210 nm min. 25 %
Transmittance at 230 nm min. 85 %
Transmittance from 254 nm min. 99 %
HPLC Gradient at 210 nm max. 50 mAU
HPLC Gradient at 254 nm max. 10 mAU
LC-MS application complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR