Acetonitrile ≥99.9% (by GC), CHROMASOLV™ NMR for LC-NMR, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34955-6X1L
BJ34955-1LCS 1675.34 USD
BJ34955-1L BJ34955-500ML
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ NMR for LC-NMR, Riedel-de Haën™
Acetonitrile

  • Solvents for LC-NMR applications

Formula: H₃CCN
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071

Order Now

Specification Test Results

Autoignition Temperature 973 °F
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0002 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0001 %
Suitability for the LC-NMR complying
Propionitrile (GC) Max. 0.003 %
Identity (NMR) complying
Vapor Density 1.41 (vs air)
Explosion Limit 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR