Potassium bromide for IR spectrophotometry, Fluka™

Supplier: Honeywell Research Chemicals

34919-6X100G
BJ34919-100GCS 2825 USD
BJ34919-100G
Potassium bromide for IR spectrophotometry, Fluka™
Potassium bromide
Formula: KBr
Boiling Pt: 1.383 °C (1.013 hPa)
Melting Pt: 734 °C
Density: 2.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3

Order Now

Specification Test Results

Suitability for IR spectroscopy complying
Vapor Pressure 1 mmHg ( 795 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR