Methanol ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34898-4X2.5L
BJ34898-2.5LCS 380.71 USD
BJ34898-2.5L
Methanol ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
Methanol

  • Solvents designed for high-performance thin-layer chromatography

Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCOOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0005 %
Transmittance at 210 nm Min. 30 %
Transmittance at 220 nm Min. 50 %
Transmittance at 235 nm Min. 80 %
Transmittance from 260 nm Min. 98 %
HPLC Gradient at 230 nm Max. 2 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
for AMD technic complying
Explosion Limit 36 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR