Acetonitrile ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34896-6X1L
BJ34896-1LCS 1180.82 USD
BJ34896-1L
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
Acetonitrile

  • Solvents designed for high-performance thin-layer chromatography

Formula: H₃CCN
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 1.41 (vs air)
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0001 %
Transmittance at 195 nm Min. 75 %
Transmittance at 200 nm Min. 93 %
Transmittance from 230 nm Min. 99 %
Baseline drift at 210 nm Max. 15 mAU
HPLC Gradient at 210 nm Max. 3 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
for AMD technic complying
Vapor Pressure 72.8 mmHg ( 20 °C)
Explosion Limit 16 %
Refractive Index n20/D 1.344 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR