Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

34885-4X4L 34885-2.5L 34885-4X2.5L 34885-200L
BJ34885-4LCS 834.59 USD
BJ34885-4L BJ34885-1L BJ34885-2.5L BJ34885-4X2.5L BJ34885-200L
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Methanol

Methanol (CH3OH) is also known as Methyl Alcohol or wood alcohol. It is a clear, colorless, volatile and flammable liquid. Methanol is a common laboratory solvent and is especially useful for HPLC and UV/VIS spectroscopy applications due to its low UV cutoff. Methanol is toxic if ingested and can result in blindness, coma and death. Suitable for HPLC gradient elution technics.


  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline


Beilstein Number: 1098229

Formula: CH₃OH
Boiling Pt: 64…65 °C (1.013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Free alkali (as NH₃) Max. 0.0005 %
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 240 nm Max. 0.05
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.15
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 0.50
HPLC Gradient at 230 nm Max. 2 mAU
HPLC Gradient at 254 nm Max. 5 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Appearance (color) complying
Appearance (form) complying
Vapor Density 1.11 (vs air)
Vapor Pressure 410 mmHg ( 50 °C)
Autoignition Temperature 725 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR