Dimethyl sulfoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning

34869-4X4L 34869-6X1L 34869-4X2.5L
BJ34869-4LCS 2802.65 USD
BJ34869-4L BJ34869-1L BJ34869-100ML BJ34869-2.5L BJ34869-2L
Dimethyl sulfoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Dimethyl sulphoxide
Formula: (CH₃)₂SO
Boiling Pt: 189 °C
Melting Pt: 18.5 °C
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 28.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.70 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.2 %
Absorbance at 350 nm Max. 0.01
Absorbance at 300 nm Max. 0.10
Absorbance 280 nm Max. 0.30
Absorbance at 270 nm Max. 0.70
Vapor Pressure 0.42 mmHg ( 20 °C)
Explosion Limit 42 %, 63 °F
Autoignition Temperature 573 °F
Vapor Density 2.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR