1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

34867-6X1L 34867-2.5L 34867-4X2.5L 34867-4X4L 34867P-4X4L
BJ34867-1LCS 904.11 USD
BJ34867-1L BJ34867-2.5L BJ34867-4X2.5L BJ34867-4X4L BJ34867P-4X4L
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
1-Butanol

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Beilstein Number: 969148
RTECS Number: 200-751-6
UNSPSC Code: EO1400000

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 9.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 99.70 %
APHA Max. 10
Non-volatile matter Max. 0.001 %
IR spectrum complying
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₃H₇COOH) Max. 0.002 %
Absorbance at 400 nm Max. 0.01
Absorbance 280 nm Max. 0.01
Absorbance at 260 nm Max. 0.04
Absorbance at 240 nm Max. 0.10
Absorbance at 235 nm Max. 0.10
Absorbance at 225 nm Max. 0.50
Absorbance at 220 nm Max. 0.70
Absorbance at 210 nm Max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR