Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

34851-4X4L 34851-200L 34851-56L 34851-4X2.5L
BJ34851-4LCS 1848.61 USD
BJ34851-4L BJ34851-1L BJ34851-2.5L BJ34851-200L BJ34851-56L BJ34851-4X2.5L
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Acetonitrile

Acetonitrile is a medium-polarity solvent that is miscible with water and a range of organic solvents, but not saturated hydrocarbons. CHROMASOLV® Gradient Grade Acetonitrile is suitable for: HPLC, TLC, GPC, UV and spectrophotometry.


  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline


Beilstein Number: 741857

Formula: H₃CCN
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7857 g/cm³ (20 °C) (lit.)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.90 %
Refractive index (n 20/D) 1.3435 - 1.3445
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0002 %
Absorbance at 400 nm Max. 0.0044
Absorbance at 250 nm Max. 0.0044
Absorbance at 235 nm Max. 0.0044
Absorbance at 230 nm Max. 0.0044
Absorbance at 200 nm Max. 0.032
Absorbance at 195 nm Max. 0.12
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
HPLC Gradient at 210 nm Max. 3 mAU
HPLC Gradient at 254 nm Max. 0.5 mAU
Baseline drift at 210 nm Max. 15 mAU
Refractive Index n20/D 1.344 (lit.)
Measuring Range ≤15 mAU baseline drift, (210 nm)
Autoignition Temperature 973 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR