Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

34850-4X4L 34850-6X1L 34850-2.5L 34850-4X2.5L
BJ34850-4LCS 884.51 USD
BJ34850-4L BJ34850-1L BJ34850-2.5L BJ34850-4X2.5L
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Acetone
Formula: CH₃COCH₃
Boiling Pt: 55…57 °C (1.013 hPa)
Melting Pt: –95 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.002 %
Transmittance at 330 nm Min. 15 %
Transmittance at 335 nm Min. 50 %
Transmittance at 340 nm Min. 80 %
Transmittance from 350 nm Min. 98 %
Vapor Density 2 (vs air)
Vapor Pressure 184 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR