1,2,4-Trichlorobenzene ≥98.0%, B&J Brand™ for GPC (gel permeation chromatography), for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Warning

348-4
BJ348-4CS 2905.34 USD
BJ348-4
1,2,4-Trichlorobenzene ≥98.0%, B&J Brand™ for GPC (gel permeation chromatography), for gas chromatography, for spectrophotometry, Burdick & Jackson™
1,2,4-Trichlorobenzene

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 213.5 °C
Melting Pt: 17 °C
Density: 1.454 g/cm³ (20 °C)
Flash Pt: 6.6 %(V) (150 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III
Merck Index: 13,09704

Order Now

Specification Test Results

GC Analysis 98 % min.
Refractive Index (20ºC) 1.5711 - 1.5721
UV Absorbance @ 310 nm 0.5 AU max.
UV Absorbance @ 350 nm 0.05 AU max.
UV Absorbance @ 375 nm 0.01 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 308 nm max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR