Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Carbamaldehyde

34724-6X1L
BJ34724-1LCS 539 USD
BJ34724-1L
Formamide, dry, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Formamide
Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1.013 hPa)
Melting Pt: 1…3 °C
Density: 1.133 g/cm³ (20 °C)
Flash Pt: 175 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
ADR: 1,
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability (for KF-titration) test passed
Water (Karl Fischer) Max. 0.02 %
Number Of Samples Min. 2
Vapor Density 1.55 (vs air)
Refractive Index n20/D 1.447 (lit.)
Vapor Pressure 0.08 mmHg ( 20 °C)
Autoignition Temperature 932 °F

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR