Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

34675-4X4L
BJ34675-4X4LCS 706 USD
BJ34675-4X4L
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Water with 0,1% (v/v) acetic acid

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9982 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Assay of acetic acid 0.093 - 0.107 %
pH (20 °C) 3.2 - 3.4
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm
Transmittance at 210 nm Min. 20 %
Transmittance at 230 nm Min. 75 %
Transmittance from 254 nm Min. 99 %
HPLC Gradient at 210 nm Max. 50 mAU
HPLC Gradient at 254 nm Max. 10 mAU
LC-MS application complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR