Water with 0,1% ammonium acetate, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

34674-4X4L
BJ34674-4X4LCS 722 USD
BJ34674-4X4L
Water with 0,1% ammonium acetate, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Water with 0,1% ammonium acetate

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Boiling Pt: 100 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 631-61-8
EINECS: 231-791-2

Order Now

Specification Test Results

Assay of ammonium acetate 0.093 - 0.107 %
pH (20 °C) 6.8 - 7.2
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm
Transmittance at 210 nm Min. 20 %
Transmittance at 230 nm Min. 90 %
Transmittance from 254 nm Min. 99 %
HPLC Gradient at 210 nm Max. 50 mAU
HPLC Gradient at 254 nm Max. 10 mAU
LC-MS application complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR