Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34671-4X2.5L 34671-4X4L
BJ34671-2.5LEA 124.18 USD
BJ34671-2.5L BJ34671-4L BJ34671-4X2.5L BJ34671-4X4L
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Methanol with 0,1% (v/v) formic acid
Boiling Pt: 64.7 °C (main component)
Melting Pt: –97.68 °C (main component)
Density: 0.792 g/cm³ (20 °C) (main component)
Flash Pt: 11 °C
Storage Temperature: Ambient
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 10 %
HPLC Gradient at 254 nm Max. 10 mAU
LC-MS application complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR