Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34668-4X2.5L 34668-4X4L
BJ34668-2.5LEA 412.83 USD
BJ34668-2.5L BJ34668-4X2.5L BJ34668-4X4L
Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Acetonitrile with 0,1% (v/v) Formic acid

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7767 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Assay of formic acid 0.093 - 0.107 %
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 15 %
Transmittance at 210 nm Min. 5 %
Suitability for the LC-MS complying
HPLC Gradient at 254 nm Max. 50 mAU
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR