Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

346136-1L 346136-6X1L
BJ346136-1LEA 197 USD
BJ346136-1L BJ346136-6X1L
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Diethyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1696894

Formula: (CH₃CH₂)₂O
Boiling Pt: 34…35 °C (1.013 hPa)
Melting Pt: –116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: –45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Titrable Acid Max. 0.0002 meq/g
Carbonyl compounds (as CO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 1 ppm
APHA Max. 10
Assay BHT (GC) Min. 1 ppm
Refractive Index n20/D 1.3530 (lit.)
Explosion Limit 36.5 %
Contains BHT as inhibitor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR