N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

34440-2.5L
BJ34440-2.5LEA 247 USD
BJ34440-2.5L
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
N,N-Dimethylformamide

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.


Beilstein Number: 605365

Formula: HCON(CH₃)₂
Boiling Pt: 152…153 °C (1.013 hPa)
Melting Pt: –61 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Water (Karl Fischer) Max. 0.02 %
Non-volatile matter Max. 0.001 %
Amines (as dimethylamine) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR