Mercury(II) nitrate 0.05 M Hg(NO3)2 (0.1N) ≥99.5% volumetric solution, for chloride determination, Fluka™

Supplier: Honeywell Research Chemicals
Danger

34292-1L
BJ34292-1LEA 191.17 USD
BJ34292-1L
Mercury(II) nitrate 0.05 M Hg(NO3)2 (0.1N) ≥99.5% volumetric solution, for chloride determination, Fluka™
Mercury (II) nitrate
Formula: HgN₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00011038
CAS Number: 10045-94-0
UN: 2024
ADR: 6.1,III

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR