Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™

Supplier: Honeywell Research Chemicals
Warning

34289-1L
BJ34289-1LEA 177.4 USD
BJ34289-1L
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Mercury (II) nitrate
Formula: HgN₂O₆
Boiling Pt: 100 °C (solvent)
Melting Pt: <0 °C
Density: 1.0 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00011038
CAS Number: 10045-94-0

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR