Oxalic acid 0.1 M (COOH)2 (0.2N) ≥98% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Ethanedioic acid

34287-1L
BJ34287-1LEA 176.1 USD
BJ34287-1L
Oxalic acid 0.1 M (COOH)2 (0.2N) ≥98% volumetric solution, Fluka™
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.999 - 1.001

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR