Potassium dichromate 1/6 M K2Cr2O7 (1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

34271-1L
BJ34271-1LEA 179.14 USD
BJ34271-1L
Potassium dichromate 1/6 M K2Cr2O7 (1N) volumetric solution, Fluka™
Potassium dichromate
Formula: K₂Cr₂O₇
Boiling Pt: 100 °C (1.013 hPa)
Density: 1.030 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR