Potassium bromate 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

34268-1L
BJ34268-1LEA 95.89 USD
BJ34268-1L
Potassium bromate 1/60 M KBrO3 (0.1N) volumetric solution, Fluka™
Potassium bromate
Formula: KBrO₃
MW: 167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2

Order Now

Specification Test Results

Assay 0.01663 - 0.01670 mol/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR