Acetic acid 1.0 M CH3COOH (1.0N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

34256-1L
BJ34256-10L-VPEA 1483.24 USD
BJ34256-10L-VP BJ34256-1L
Acetic acid 1.0 M CH3COOH (1.0N) volumetric solution, Fluka™
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 100 °C
Density: 1.000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR