Ammonium iron (III) sulfate 0.1 M (NH4)Fe(SO4)2 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

34249-1L
BJ34249-1LEA 174.71 USD
BJ34249-1L
Ammonium iron (III) sulfate 0.1 M (NH4)Fe(SO4)2 volumetric solution, Fluka™
Ammonium iron (III) sulphate
Formula: H₄FeNO₈S₂
MDL Number: MFCD00012019
CAS Number: 10138-04-2

Order Now

Specification Test Results

Factor 0.998 - 1.002

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR