Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Diameter ~2 mm, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

34241-250G
BJ34241-250GEA 99 USD
BJ34241-250G
Molecular sieve 3A (0.3 nm, 3 Å), Hydranal™ drying agent, for air and gases for Karl Fischer titration, Diameter ~2 mm, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Density: 2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Specification Test Results

Suitability (for KF-titration) test passed

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR