4-Methyl-2-pentanone ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur. for extraction analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

33463-1L 33463-2.5L 33463-4X2.5L
BJ33463-1LEA 128.66 USD
BJ33463-1L BJ33463-2.5L BJ33463-4X2.5L
4-Methyl-2-pentanone ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur. for extraction analysis, Riedel-de Haën™
4-Methyl-2-pentanone

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 605399

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 115…117 °C (1.013 hPa)
Melting Pt: –83 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
UN: 1245
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Non-volatile matter Max. 10 ppm
Water (Karl Fischer) Max. 500 ppm
Titrable Acid Max. 0.002 meq/g
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Calcium (Ca) Max. 0.5 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
APHA Max. 15
Appearance complying
Explosion Limit 1.2-8 %, 93 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR