Molecular sieve A5 (0.5 nm, 5 Å), pellets, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets

334316-1KG
BJ334316-1KGEA 238 USD
BJ334316-1KG
Molecular sieve A5 (0.5 nm, 5 Å), pellets, Fluka™
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now

Specification Test Results

Particle Size 1.6 mm
Form pellets
Granulation complying
Pore Size (Ång Ström) 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR