Methyl red solution in ethanol acid-base indicator, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

32941-100ML
BJ32941-100MLEA 87.3 USD
BJ32941-100ML
Methyl red solution in ethanol acid-base indicator, Fluka™
Methyl red
Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

Change complying
Test of sensitivity complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR