Bromothymol blue solution in ethanol acid-base indicator, Fluka™

Supplier: Honeywell Research Chemicals
Warning

32822-100ML
BJ32822-100MLEA 87.19 USD
BJ32822-100ML
Bromothymol blue solution in ethanol acid-base indicator, Fluka™
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

Test of sensitivity complying
Change range (pH 5,8-7,4) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR