Eriochrome Black T Reag. Ph. Eur. indicator for metal titration, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

32751-100G
BJ32751-100GEA 101.21 USD
BJ32751-100G
Eriochrome Black T Reag. Ph. Eur. indicator for metal titration, Fluka™
Eriochrome Black T
Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3

Order Now

Specification Test Results

Identity (IR) complying
Solubility in methanol complying
Indicator solution complying
Suitability for Complexometry complying
UV spectrum complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR