Bromocresol green Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32742-5G
BJ32742-5GEA 94.11 USD
BJ32742-5G
Bromocresol green Reag. Ph. Eur., Fluka™
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH Range 3.8 - 5.4, yellow to blue
Indicator solution complying
Change range complying
Test of sensitivity complying
Curve course (UV) complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR