Phenol red ACS indicator, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
32661-100G
32661-10G
32661-25G
BJ32661-100GEA
407.1
USD
BJ32661-100G
BJ32661-10G
BJ32661-25G
Phenol red ACS indicator, Fluka™
Phenol red
Formula:
C₁₉H₁₄O₅S Storage Temperature: Ambient |
MDL Number:
MFCD00003552 CAS Number: 143-74-8 EINECS: 205-609-7 |
Specification Test Results
Clearness of the solution | complying |
Loss on drying (vacuum) | Max. 5 % |
Residue on ignition | Max. 1 % |
Change range | complying |
Extinction maximum at | 552 - 562 nm |
Extinction maximum of | 900 - 1800 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...