Cresol red ≥83% indicator, Fluka™

Supplier: Honeywell Research Chemicals

32653-25G
BJ32653-10GEA 68.49 USD
BJ32653-10G BJ32653-25G
Cresol red ≥83% indicator, Fluka™
Cresol red
Formula: C₂₁H₁₈O₅S
Density: 0.998 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005878
CAS Number: 1733-12-6
EINECS: 217-064-2

Order Now

Specification Test Results

Identity (IR) complying
Assay (HPLC) Min. 83 %
Loss on drying Max. 5 %
Change range (pH 0.5-2.5) complying
Change range (pH 6.5-8.5) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR