Fluorescein free acid Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals

32615-100G
BJ32615-100GEA 88.32 USD
BJ32615-100G BJ32615-25G
Fluorescein free acid Reag. Ph. Eur., Fluka™
Fluorescein
Formula: C₂₀H₁₂O₅
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00005050
CAS Number: 2321-07-5
EINECS: 219-031-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b9d8d37f0106f4c8ef2d7759759d21bd43cc03&vid=25b78df95a04ab26c5ba8c96b95f031056926b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ignition Residue ≤0.25% (as SO₄)
Identity (IR) complying
UV spectrum complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR