Sodium acetate, anhydrous ≥99.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals

32319-6X1KG
BJ32319-1KGCS 1045.42 USD
BJ32319-1KG BJ32319-500G
Sodium acetate, anhydrous ≥99.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: Decomposes on heating
Melting Pt: 324 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8

Order Now

Specification Test Results

Assay (HClO₄-titr.) Min. 99.0 %
Water insoluble matter Max. 0.005 %
Loss on drying (120°C) Max. 1.0 %
pH (5 %, 25 °C) 7.0 - 9.2
Arsenic (As) Max. 0.00001 %
Aluminium (Al) Max. 0.001 %
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.02 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.0005 %
Sulfate (SO₄) Max. 0.002 %
Appearance of the solution complying
Colour of the solution complying
Turbidity of solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR