Nitric acid concentrate for dilution to 1L, 0.1 M HNO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

32042-1EA
BJ32042-1EAEA 87.9 USD
BJ32042-1EA BJ32042-6X1EA
Nitric acid concentrate for dilution to 1L, 0.1 M HNO3 (0.1N) volumetric solution, Fluka™
Nitric acid

For 1L standard solution

Formula: HNO₃
Boiling Pt: 100 °C
Melting Pt: no data available
Density: 1.065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR