Fluka™ Fluoride buffer solution According to DEV D4, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
32028-1L
BJ32028-1LEA 126 USD
BJ32028-1L
Fluka™ Fluoride buffer solution According to DEV D4, Honeywell Research Chemicals
Buffers Ionic Strength Adjustment Buffers
TISAB according to DEV D4.