Starch 1% in water stabilized indicator, Fluka™

Supplier: Honeywell Research Chemicals

319554-500ML
BJ319554-500MLEA 470.62 USD
BJ319554-500ML
Starch 1% in water stabilized indicator, Fluka™
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.005 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now

Specification Test Results

Turbidity Max. 50 NTU
Test of sensitivity complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR