Sodium hydroxide 1.0 M NaOH (1.0N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
319511-1L
319511-2L
319511-500ML
319511-6X1L
BJ319511-1LEA
36
USD
BJ319511-1L
BJ319511-2L
BJ319511-500ML
BJ319511-6X1L
Sodium hydroxide 1.0 M NaOH (1.0N) volumetric solution, Fluka™
Sodium hydroxide
Formula:
NaOH Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,II |
Specification Test Results
Assay | 0.950 - 1.050 N |
Carbonate (as Na₂CO₃) | Max. 0.03 N |
Traceable to NIST SRM | effective |
Vapor Pressure | 3 mmHg ( 37 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...