Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

319481-1L 319481-2.5L 319481-2L 319481-500ML 319481-6X500ML
BJ319481-1LEA 81 USD
BJ319481-1L BJ319481-2.5L BJ319481-2L BJ319481-500ML BJ3194816500ML
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Assay titr. 0.095 - 0.105 mol/l
Carbonate (CO₃) Max. 0.0015 mol/l
Traceable to NIST SRM effective
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR