Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

319376-2L
BJ319376-2LEA 108.56 USD
BJ319376-2L BJ319376-500ML
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Potassium hydroxide
Formula: KOH
Boiling Pt: 100 °C
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance complying
Coloration complying
Clearness complying
Assay titr. 0.950 - 1.050 mol/l
Traceable to NIST SRM complying
BAM/EMPA reference material complying
BAM/EMPA Certificate Number complying
Carbonate (CO₃) Max. 0.01 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR