Hydrochloric acid 0.5 M HCl (0.5N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

318957-2L
BJ318957-2LEA 136 USD
BJ318957-2L BJ318957-500ML
Hydrochloric acid 0.5 M HCl (0.5N) volumetric solution, Fluka™
Hydrochloric acid
Formula: HCl
Boiling Pt: >100 °C (1.013 hPa)
Melting Pt: 0 °C
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Assay 0.4900 - 0.5100 N
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR