Sodium peroxide ≥95%, Puriss. p.a. ACS, ISO, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: di-Sodium peroxide

31445-6X1KG
BJ31445-100GEA 258.34 USD
BJ31445-100G BJ31445-1KG BJ31445-500G
Sodium peroxide ≥95%, Puriss. p.a. ACS, ISO, Fluka™
Sodium peroxide
Formula: Na₂O₂
MW: 77.98 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 660 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I

Order Now

Specification Test Results

Assay Min. 95.0 %
Aluminium (Al) Max. 0.001 %
Iron (Fe) Max. 0.002 %
Heavy metals (as Pb) Max. 0.002 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.001 %
Total N Max. 0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR